Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNG2 Monoclonal Antibody (7C13)
GREENER_CHOICE

Catalog No. PIMA549295
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549295 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549295 Supplier Invitrogen™ Supplier No. MA549295
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue. ICC/IF: Caco-2 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This gene encodes one of the gamma subunits of a guanine nucleotide-binding protein. Such proteins are involved in signaling mechanisms across membranes. Various subunits forms heterodimers which then interact with the different signal molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNG2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 7C13
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene GNG2
Gene Accession No. P59768, P63213
Gene Alias 82; G gamma-I; G protein subunit gamma 2; GBG2; GNG2; guanine nucleotide binding protein (G protein), gamma 2; guanine nucleotide binding protein (G protein), gamma 2 subunit; guanine nucleotide binding protein gamma 2; guanine nucleotide-binding protein G(I)/G(O) gamma-2 subunit; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-2
Gene Symbols GNG2
Host Species Mouse
Immunogen E.coli-derived human GNG2 recombinant protein (Position: A2-D48).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14702, 54331, 80850
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.