Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551954 Supplier Invitrogen™ Supplier No. PA551954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASIAQARKLV EQLKMEANID RIKVSKAAAD LMAYCEAHAK EDPLLTPVPA SENPFREK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 79%.

This gene encodes one of the gamma subunits of a guanine nucleotide-binding protein. Such proteins are involved in signaling mechanisms across membranes. Various subunits forms heterodimers which then interact with the different signal molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNG2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GNG2
Gene Accession No. P59768
Gene Alias 82; G gamma-I; G protein subunit gamma 2; GBG2; GNG2; guanine nucleotide binding protein (G protein), gamma 2; guanine nucleotide binding protein (G protein), gamma 2 subunit; guanine nucleotide binding protein gamma 2; guanine nucleotide-binding protein G(I)/G(O) gamma-2 subunit; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-2
Gene Symbols GNG2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GNG2. Recombinant protein control fragment (Product #RP-89145).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54331
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.