Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNG3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576438
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Specifications
GNG3 | |
Polyclonal | |
Unconjugated | |
GNG3 | |
[g]3; G protein subunit gamma 3; Gng3; Gngt3; G-protein gamma 3 subunit; guanine nucleotide binding protein (G protein), gamma 3; guanine nucleotide binding protein (G protein), gamma 3 subunit; guanine nucleotide binding protein, gamma 3; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-3; guanine nucleotide-binding protein gamma-3 subunit; guanine nucleotide-binding regulatory protein gamma-3 subunit; NBP gamma-3; wu:fj56b02; zgc:92852 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114117, 14704, 2785 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P63215, P63216 | |
GNG3 | |
Recombinant protein of human GNG3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction