Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597256
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597256 Supplier Invitrogen™ Supplier No. PA597256
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKGETPVNST MSIGQARKMV EQLKIEASLC RIKVSKAAAD LMTYCDAHAC EDPLITPVPT SENPFREKKF FCALL; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell membrane, Cytoplasmic side, Lipid-anchor.

Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNG3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GNG3
Gene Accession No. P63216
Gene Alias [g]3; G protein subunit gamma 3; Gng3; Gngt3; G-protein gamma 3 subunit; guanine nucleotide binding protein (G protein), gamma 3; guanine nucleotide binding protein (G protein), gamma 3 subunit; guanine nucleotide binding protein, gamma 3; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-3; guanine nucleotide-binding protein gamma-3 subunit; guanine nucleotide-binding regulatory protein gamma-3 subunit; NBP gamma-3; wu:fj56b02; zgc:92852
Gene Symbols GNG3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-75 of human GNG3 (NP_0363341).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114117, 14704
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.