Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570824
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Specifications
GNG4 | |
Polyclonal | |
Unconjugated | |
GNG4 | |
G protein subunit gamma 4; Gng4; GNGT4; guanine nucleotide binding protein (G protein), gamma 4; guanine nucleotide binding protein (G protein), gamma 4 subunit; guanine nucleotide binding protein 4; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2786 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P50150 | |
GNG4 | |
synthetic peptide directed towards the middle region of human GNG4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction