Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119901
This item is not returnable.
View return policy
Description
Positive Control: SiHa, human pancreas tissue, human kidney tissue, mouse kidney tissue. Subcellular Location: Cell membrane.
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Specifications
| GNG4 | |
| Polyclonal | |
| Unconjugated | |
| GNG4 | |
| G protein subunit gamma 4; Gng4; GNGT4; guanine nucleotide binding protein (G protein), gamma 4; guanine nucleotide binding protein (G protein), gamma 4 subunit; guanine nucleotide binding protein 4; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14706, 2786 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P50150, P50153 | |
| GNG4 | |
| Recombinant protein within human GNG4 aa 1-75. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction