Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNG7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568206
Description
Predicted to react with Mouse and Rat samples.
GNG7 is a Guanine nucleotide-binding protein (G proteins) and are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction. Plays a role in the regulation of adenylyl cyclase signaling in certain regions of the brain. Plays a role in the formation or stabilization of a G protein heterotrimer (G(olf) subunit alpha-beta-gamma-7) that is required for adenylyl cyclase activity in the striatum.
Specifications
GNG7 | |
Polyclonal | |
Unconjugated | |
Gng7 | |
AI840417; G protein subunit gamma 7; GNG7; Gngt7; guanine nucleotide binding protein (G protein), gamma 7; guanine nucleotide binding protein (G protein), gamma 7 subunit; guanine nucleotide binding protein, gamma 7; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-7 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14708, 2788, 58979 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O60262, P43425, Q61016 | |
Gng7 | |
Synthesized peptide derived from human GNG7. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction