Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNG7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563485
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563485 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563485 Supplier Thermo Scientific Supplier No. PA563485
Add to Cart
Add to Cart

GNG7 Polyclonal Antibody

GNG7 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen GNG7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gng7
Gene Accession No. O60262
Gene Alias AI840417; G protein subunit gamma 7; GNG7; Gngt7; guanine nucleotide binding protein (G protein), gamma 7; guanine nucleotide binding protein (G protein), gamma 7 subunit; guanine nucleotide binding protein, gamma 7; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-7
Gene Symbols Gng7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GNG7. Recombinant protein control fragment (Product #RP-104516).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2788
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.