Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNGT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549350
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549350 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549350 Supplier Invitrogen™ Supplier No. PA549350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with bovine based on sequence homology.

Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein- effector interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNGT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Gngt1
Gene Accession No. P63211, Q61012
Gene Alias G protein subunit gamma transducin 1; G(y)1; Gng1; Gngt1; guanine nucleotide binding protein (G protein), gamma 1 subunit; guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 1; guanine nucleotide binding protein, gamma transducing activity polypeptide 1; guanine nucleotide-binding protein G(T) subunit gamma-T1; Transducin gamma chain
Gene Symbols Gngt1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide within amino acid positions 54-74 of human GNGT1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14699, 2792
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.