Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNGT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589564
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589564 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589564 Supplier Invitrogen™ Supplier No. PA589564
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAQDLSEKDL LKMEVEQLKK EVKNTRIPIS KAGKEIKEYV EAQAGNDPFL KGIPEDKNPF KEKGGCLIS; Positive Samples: Mouse spleen; Cellular Location: Cell membrane, Cytoplasmic side, Lipid-anchor.

Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNGT2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GNGT2
Gene Accession No. O14610, Q61017
Gene Alias AV096488; G gamma-C; G protein cone gamma 8 subunit; G protein subunit gamma transducin 2; gamma-T2 subunit; G-gamma-8; G-gamma-9; G-GAMMA-C; GNG8; GNG9; Gngt2; Gngt8; guanidine nucleotide binding protein, (G protein) , gamma transducing activity polypeptide 1; guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2; guanine nucleotide binding protein (G protein), gamma transducing polypeptide 2; guanine nucleotide binding protein gamma 9; guanine nucleotide binding protein gamma transducing activity polypeptide 2; guanine nucleotide binding protein, gamma transducing activity polypeptide 2; guanine nucleotide regulatory protein gamma c subunit; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-T2; Tgamma c; transducin gamma subunit
Gene Symbols GNGT2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-69 of human GNGT2 (NP_1136861).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14710, 2793
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.