Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101628
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101628 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101628 Supplier Thermo Scientific Supplier No. PA5101628
Add to Cart
Add to Cart

GNL1 Polyclonal Antibody

GNL1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen GNL1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GNL1
Gene Accession No. P36915, P36916, Q6MG06
Gene Alias G protein nucleolar 1 (putative); Gnal1; Gna-rs1; GNL1; GTP-binding protein HSR1; GTP-binding protein MMR1; guanine nucleotide binding protein, related sequence 1; guanine nucleotide binding protein-like 1; guanine nucleotide-binding protein-like 1; HSR1; HSR1 GTP-binding protein; Mmr1
Gene Symbols GNL1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GNL1(Accession P36915), corresponding to amino acid residues V9-H59.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14670, 2794, 309593
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.