Learn More
GNL1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | GNL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-2129
![]() |
Bethyl Laboratories, Inc
A302247AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Orangutan, Monkey and Chimpanzee
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.Specifications
GNL1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
HSR1, GTP-binding protein HSR1, HSR1 GTP-binding protein, guanine nucleotide-binding protein-like 1 | |
GNL1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P36915 | |
2794 | |
Between 50 and 100 | |
Primary | |
4° C | |
GNL1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.