Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18164925UL
Description
GNL2 Polyclonal specifically detects GNL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GNL2 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Autoantigen NGP-1, FLJ40906, guanine nucleotide binding protein-like 2 (nucleolar), HUMAUANTIG, Ngp-1, NGP1novel nucleolar guanosine 5'-triphosphate binding protein autoantigen, nucleolar G-protein gene 1, nucleolar GTPase, nucleolar GTP-binding protein 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
29889 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
GNL2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:RIPFFVKPPNAEPLVAPQLLPSSSLEVVPEAAQNNPGEEVTETAGEGSESIIKEETEENSHCDANTEMQQILTRVRQNFGKINVVPQFSGDDLVPV | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction