Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589063
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589063 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589063 Supplier Invitrogen™ Supplier No. PA589063
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MVKPKYKGRS TINPSKASTN PDRVQGAGGQ NMRDRATIRR LNMYRQKERR NSRGKIIKPL QYQSTVASGT VARVEPNIKW FGNTRVIKQS SLQKFQEEMD TVMKDPYKVV MKQSKLPMSL LHDRIRPHNL KVHILDTESF ETTFGPKSQR KRPNLFASDM QSLIENAEMS TESYDQGKDR DLVTEDTGVR NEAQEEIYKK GQSKRIWGEL YKVIDSSDVV VQVLDARDPM GTRSPHIETY LKKEKPWKHL IFVLNKCDLV PTWATKRWVA VLSQDYPTLA FHASLTNPFG KGAFIQLLRQ; Positive Samples: NIH/3T3; Cellular Location: Nucleus, nucleolus.

GTPase that associates with pre-60S ribosomal subunits in the nucleolus and is required for their nuclear export and maturation. May promote cell proliferation possibly by increasing p53/TP53 protein levels, and consequently those of its downstream product CDKN1A/p21, and decreasing RPL23A protein levels (PubMed:26203195). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GNL2
Gene Accession No. Q13823, Q99LH1
Gene Alias Autoantigen NGP-1; BC003262; G protein nucleolar 2; GNL2; guanine nucleotide binding protein-like 2 (nucleolar); HUMAUANTIG; NGP1; Ngp-1; Nog2; novel nucleolar guanosine 5'-triphosphate binding protein autoantigen; nucleolar G-protein gene 1; nucleolar GTPase; nucleolar GTP-binding protein 2; nucleostemin-2; Nug2; RGD1305006; testicular antigen
Gene Symbols GNL2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GNL2 (NP_0374171).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230737, 29889
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.