Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNL2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982892
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2892 Supplier Bioss Supplier No. BS13471R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GTPase that associates with pre-60S ribosomal subunits in the nucleolus and is required for their nuclear export and maturation.

Specifications

Antigen GNL2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GNL2
Gene Accession No. Q13823
Gene Alias Autoantigen NGP-1; BC003262; G protein nucleolar 2; GNL2; guanine nucleotide binding protein-like 2 (nucleolar); HUMAUANTIG; NGP1; Ngp-1; Nog2; novel nucleolar guanosine 5′-triphosphate binding protein autoantigen; nucleolar G-protein gene 1; nucleolar GTPase; Nucleolar GTP-binding protein 2; nucleostemin-2; Nug2; testicular antigen
Gene Symbols GNL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GNL2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29889
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.