Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNL3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544940 Supplier Invitrogen™ Supplier No. PA544940
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MMKLRHKNKK PGEGSKGHKK ISWPYPQPAK QNGKKATSKV PSAPHFVHPN Sequence homology: Cow: 86%; Dog: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rabbit: 86%; Rat: 86%.

The protein encoded by this gene appears to be a nucleolar GTPase that is essential for ribosomal pre-rRNA processing and cell proliferation. Two transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNL3L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GNL3L
Gene Accession No. Q9NVN8
Gene Alias BC020354; G protein nucleolar 3 like; G protein nucleolar 3B; GNL3B; GNL3L; guanine nucleotide binding protein-like 3 (nucleolar)-like; guanine nucleotide-binding protein-like 3-like protein; hypothetical protein LOC507679; novel GTPase
Gene Symbols GNL3L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GNL3L (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54552
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.