Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GNL3L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583321
Description
Immunogen sequence: QPAKQNGKKA TSKVPSAPHF VHPNDHANRE AELKKKWVEE MREKQQAARE QERQKRRTIE SYCQDVLRRQ EEFEHKEEVL QELN.
The protein encoded by this gene appears to be a nucleolar GTPase that is essential for ribosomal pre-rRNA processing and cell proliferation. Two transcript variants encoding the same protein have been found for this gene.
Specifications
| GNL3L | |
| Polyclonal | |
| Unconjugated | |
| GNL3L | |
| BC020354; G protein nucleolar 3 like; G protein nucleolar 3B; GNL3B; GNL3L; guanine nucleotide binding protein-like 3 (nucleolar)-like; guanine nucleotide-binding protein-like 3-like protein; hypothetical protein LOC507679; novel GTPase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54552 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NVN8 | |
| GNL3L | |
| Recombinant protein corresponding to Human GNL3L. Recombinant protein control fragment (Product #RP-95461). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction