Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555637
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555637 Supplier Thermo Scientific Supplier No. PA555637
Add to Cart
Add to Cart

GNMT Polyclonal Antibody

GNMT Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen GNMT
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GNMT
Gene Accession No. Q14749
Gene Alias 2.1.1.20; epididymis secretory sperm binding protein Li 182mP; folate-binding protein; glycine methyltransferase; glycine N methyl transferase; glycine N-methyltransferase; GNMT; HEL-S-182mP
Gene Symbols GNMT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GNMT. Recombinant protein control fragment (Product #RP-94803).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27232
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.