Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNMT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA576962
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA576962 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA576962 Supplier Invitrogen™ Supplier No. PA576962
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

The protein encoded by this gene is an enzyme that catalyzes the conversion of S-adenosyl-L-methionine to S-adenosyl-L-homocysteine and sarcosine. The encoded protein is found in the cytoplasm and acts as a homotetramer. Defects in this gene are a cause of GNMT deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNMT
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene GNMT
Gene Accession No. P13255, Q14749, Q9QXF8
Gene Alias 2.1.1.20; epididymis secretory sperm binding protein Li 182mP; folate-binding protein; glycine methyltransferase; glycine N methyl transferase; glycine N-methyltransferase; GNMT; HEL-S-182mP
Gene Symbols GNMT
Host Species Rabbit
Immunogen Recombinant full length Human GNMT.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14711, 25134, 27232
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.