Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNPAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536447
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536447 Supplier Invitrogen™ Supplier No. PA536447
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 77 kDa. Purity is >95% by SDS-PAGE.

GNPAT is a key enzyme located in the peroxisomal membrane which is essential to the synthesis of ether phospholipids. Mutations in GNPAT are associated with rhizomelic chondrodysplasia punctata type 2, which is characterized by rhizomelic shortening of femur and humerus, vertebral disorders, cataract, cutaneous lesions and severe mental retardation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNPAT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene GNPAT
Gene Accession No. O15228, P98192, Q9ES71
Gene Alias acyl-CoA:dihydroxyacetonephosphate acyltransferase; acyl-CoA:dihydroxyacetonephosphateacyltransferase; AU019525; D1Ertd819e; DAPAT; DAP-AT; DHAPAT; DHAP-AT; Dihydroxyacetone phosphate acyltransferase; glycerone-phosphate O-acyltransferase; glyceronephosphate O-acyltransferase; GNPAT; RCDP2
Gene Symbols GNPAT
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 341-390 of human GNPAT.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14712, 8443, 84470
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.