Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNPAT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539135
Description
GNPAT is a key enzyme located in the peroxisomal membrane which is essential to the synthesis of ether phospholipids. Mutations in GNPAT are associated with rhizomelic chondrodysplasia punctata type 2, which is characterized by rhizomelic shortening of femur and humerus, vertebral disorders, cataract, cutaneous lesions and severe mental retardation.
Specifications
| GNPAT | |
| Polyclonal | |
| Unconjugated | |
| GNPAT | |
| Dihydroxyacetone phosphate acyltransferase, DAP-AT, DHAP-AT, Acyl-CoA:dihydroxyacetonephosphateacyltransferase, Glycerone-phosphate O-acyltransferase, GNPAT, DAPAT, DHAPAT | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8443 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15228 | |
| GNPAT | |
| A synthetic peptide derived from the C-terminal region of human GNPAT | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction