Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNPAT Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GNPAT |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GNPAT Polyclonal specifically detects GNPAT in Human, Mouse samples. It is validated for Western Blot.Specifications
GNPAT | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
8443 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Acyl-CoA:dihydroxyacetonephosphateacyltransferase, DAPATdihydroxyacetone phosphate acyltransferase, DHAP-AT, DHAPATDAP-ATacyl-CoA:dihydroxyacetonephosphateacyltransferase, EC 2.3.1, EC 2.3.1.42, glyceronephosphate O-acyltransferase, Glycerone-phosphate O-acyltransferase | |
Recombinant fusion protein containing a sequence corresponding to amino acids 481-680 of human GNPAT (NP_055051.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title