Learn More
GNPAT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 149311AP150UL

Description
This antibody recognize the 69 kDa GNPAT protein.
GNPAT is a key enzyme located in the peroxisomal membrane which is essential to the synthesis of ether phospholipids. Mutations in GNPAT are associated with rhizomelic chondrodysplasia punctata type 2, which is characterized by rhizomelic shortening of femur and humerus, vertebral disorders, cataract, cutaneous lesions and severe mental retardation.Specifications
GNPAT | |
Polyclonal | |
Unconjugated | |
GNPAT | |
DAP AT, DAPAT, DHAP AT, DHAPAT, GNPAT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14712, 8443, 84470 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15228, P98192, Q9ES71 | |
GNPAT | |
GNPAT Fusion Protein Ag6735 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.