Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNPDA1 Monoclonal Antibody (237CT2.5.2)
GREENER_CHOICE

Catalog No. PIMA537517
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537517 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537517 Supplier Invitrogen™ Supplier No. MA537517
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GNPDA1 Monoclonal Antibody for Western Blot

GNPDA1 is a 289 amino acid cytoplasmic protein belonging to the glucosamine/ galactosamine-6-phosphate isomerase family. It is one of the two mammalian glucosamine-6-phosphate deaminase enzymes. GNPDA1 is an allosteric enzyme that catalyzes the reversible conversion of D-glucosamine-6-phosphate (GlcN6P) into D-fructose-6-phosphate (Fru6P) and ammonium with an aldo/keto isomerization and an amination/deamination. GNPDA1 may trigger calcium oscillations in mammalian eggs at fertilization which in turn serve as an important factor for egg development and early embryo development. It is a homohexamer, ubiquitously expressed in most of human tissues with high expression in testis, ovary, placenta, and heart and also in various cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNPDA1
Applications Western Blot
Classification Monoclonal
Clone 237CT2.5.2
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GNPDA1
Gene Accession No. O88958
Gene Alias glcN6P deaminase 1; glucosamine-6-phosphate deaminase 1; Glucosamine-6-phosphate isomerase 1; glucose-6-phosphate isomerase; Gnp1; GNPDA; GNPDA 1; GNPDA1; Gnpi; GPI; HLN; hypothetical protein LOC550565; KIAA0060; oscillin; zgc:110691
Gene Symbols GNPDA1
Host Species Mouse
Immunogen GNPDA1 recombinant protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26384
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.