Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNPDA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GNPDA1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GNPDA1 Polyclonal specifically detects GNPDA1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GNPDA1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 3.5.99.6, GlcN6P deaminase 1, glucosamine-6-phosphate deaminase 1KIAA0060HLNGNPIGNPDA, glucosamine-6-phosphate isomerase, glucosamine-6-phosphate isomerase 1, GNP1, GNPDA 1, GPI, oscillin | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human GNPDA1 (NP_005462.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS with 50% glycerol, pH7.3. | |
10007 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title