Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNPDA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982894
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2894 Supplier Bioss Supplier No. BS13473R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GNPDA1 is a 289 amino acid cytoplasmic protein belonging to the glucosamine/ galactosamine-6-phosphate isomerase family. It is one of the two mammalian glucosamine-6-phosphate deaminase enzymes. GNPDA1 is an allosteric enzyme that catalyzes the reversible conversion of D-glucosamine-6-phosphate (GlcN6P) into D-fructose-6-phosphate (Fru6P) and ammonium with an aldo/keto isomerization and an amination/deamination. GNPDA1 may trigger calcium oscillations in mammalian eggs at fertilization which in turn serve as an important factor for egg development and early embryo development. It is a homohexamer, ubiquitously expressed in most of human tissues with high expression in testis, ovary, placenta, and heart and also in various cell lines.

Specifications

Antigen GNPDA1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GNPDA1
Gene Accession No. P46926
Gene Alias BOS_8262; glcN6P deaminase 1; Glucosamine-6-phosphate deaminase 1; Glucosamine-6-phosphate isomerase 1; glucose-6-phosphate isomerase; GNP1; GNPDA; GNPDA 1; GNPDA1; Gnpi; GPI; HLN; hypothetical protein LOC550565; KIAA0060; LOC100355853; Oscillin; zgc:110691
Gene Symbols GNPDA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GNPDA1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10007
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.