Learn More
Invitrogen™ GNPDA2 Polyclonal Antibody

Description
A suggested positive control is human kidney tissue lysate. PA5-20694 can be used with blocking peptide PEP-0809.
Specifications
Specifications
| Antigen | GNPDA2 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Gnpda2 |
| Gene Accession No. | Q8TDQ7, Q9CRC9 |
| Gene Alias | 4921523I18Rik; 4933412A11Rik; BB189630; glcN6P deaminase 2; glucosamine-6-phosphate deaminase 2; Glucosamine-6-phosphate isomerase 2; glucosamine-6-phosphate isomerase SB52; GNP2; GNPDA; GNPDA 2; GNPDA2; Kiaa4008; mKIAA4008; oscillin; Sb52 |
| Gene Symbols | Gnpda2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.