Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNPNAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100547 Supplier Invitrogen™ Supplier No. PA5100547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GNPNAT1.

Glucosamine-6 phosphate-N-acetyl Transferase (GNPNAT1) is a key enzyme in the hexosamine biosynthetic pathway. It converts D-glucosamine 6-phosphate to N-acetyl-D-glucosamine 6-phosphate, the acetylation step allowing carbon and nitrogen enter into the hexosamine biosynthetic pathway. Recently inhibition of GNPNAT1 has been reported to promote proliferation and aggressiveness of castration-resistant prostate cancer. (27194471).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNPNAT1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GNPNAT1
Gene Accession No. Q96EK6, Q9JK38
Gene Alias AU017428; AU040593; EMeg32; Glucosamine 6-phosphate N-acetyltransferase; glucosamine-6-phosphate acetyltransferase; glucosamine-phosphate N-acetyltransferase 1; glucosamine-phosphate N-acetyltransferase 1; glucosamine-6-phosphate acetyltransferase; glucosamine-phosphate N-acetyltransferase; GNA1; GNPNAT; GNPNAT1; Gpnat1; Gsnpat; Phosphoglucosamine acetylase; phosphoglucosamine transacetylase; Protein EMeg32; RGD1563144
Gene Symbols GNPNAT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GNPNAT1(Accession Q96EK6), corresponding to amino acid residues E5-K55.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498486, 54342, 64841
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.