Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNPTAB Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980890
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0890 Supplier Bioss Supplier No. BS10460R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Catalyzes the formation of mannose 6-phosphate (M6P) markers on high mannose type oligosaccharides in the Golgi apparatus. M6P residues are required to bind to the M6P receptors (MPR), which mediate the vesicular transport of lysosomal enzymes to the endosomal/prelysosomal compartment.

Specifications

Antigen GNPTAB
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene GNPTAB
Gene Accession No. Q3T906
Gene Alias BOS_5523; EG432486; GlcNAc phosphotransferase; GlcNAc-1-phosphotransferase subunits alpha/beta; glucosamine (UDP-N-acetyl)-lysosomal-enzyme N-acetylglucosamine phosphotransferase; gnpta; GNPTAB; ICD; KIAA1208; mKIAA1208; N-acetylglucosamine-1-phosphate transferase alpha and beta subunits; N-acetylglucosamine-1-phosphate transferase subunits alpha and beta; N-acetylglucosamine-1-phosphate transferase, alpha and beta subunits; N-acetylglucosamine-1-phosphotransferase subunit alpha; N-acetylglucosamine-1-phosphotransferase subunit beta; N-acetylglucosamine-1-phosphotransferase subunits alpha/beta; RGD1564821; si:ch211-234f20.3; Stealth protein gnptab; UDP-N-acetylglucosamine-1-phosphotransferase subunits alpha/beta; UDP-N-acetylglucosamine-lysosomal-enzyme N-acetylglucosamine; Unknown (protein for MGC:122985); wu:fd55d09; zgc:122985; zgc:122985 protein; zgc:158502
Gene Symbols GNPTAB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human N-acetylglucosamine-1-phosphotransferase subunit alpha.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79158
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.