Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GNPTG Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | GNPTG |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GNPTG Polyclonal specifically detects GNPTG in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GNPTG | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
84572 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PHALLVYPTLPEALQRQWDQVEQDLADELITPQGHEKLLRTLFEDAGYLKTPEENEPTQLEGGPDSLGFETLENCRKAHKELSKEIKRLKGLLTQHGIPYTRPTETSNLEHLG | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
C16orf27, c316G12.3, CAB56184, GlcNAc-1-phosphotransferase subunit gamma, GNPTAGchromosome 16 open reading frame 27, LP2537, N-acetylglucosamine-1-phosphate transferase, gamma subunit, N-acetylglucosamine-1-phosphotransferase subunit gamma, N-acetylglucosamine-1-phosphotransferase, gamma subunit, RJD9, UDP-N-acetylglucosamine-1-phosphotransferase subunit gamma | |
GNPTG | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title