Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GnRH Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 269501AP150UL

Description
The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).Specifications
GnRH | |
Polyclonal | |
Unconjugated | |
GNRH1 | |
GNRH, GnRH associated peptide I, GnRH I, GNRH1, Gonadoliberin I, Gonadorelin, GRH, LHRH, LNRH, Luliberin I, Progonadoliberin 1, Progonadoliberin I | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2796 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P01148 | |
GNRH1 | |
GnRH Fusion Protein Ag25667 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction