Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ GnRH Receptor/LH-RH Receptor Ab-3, Mouse Monoclonal Antibody

Catalog No. MS1139R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS1139R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1139R1 Supplier Epredia™ Supplier No. MS1139R1

Obtain accurate, reproducible results in immunohistochemistry, immunofluorescence and flow cytometry experiments.

GnRH receptor contains seven hydrophobic transmembrane domains connected by hydrophilic extracellular and intracellular loops characteristic of G-protein coupled receptors. GnRH stimulates the gonadotrophs of the anterior pituitary to secrete luteinizing hormone (LH) as well as follicle-stimulating hormone (FSH).

Host Species: Mouse

Clone: GNRH03; same as F1G4

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: aa 1-29

Immunogen: A synthetic peptide aa 1-29 (MAN-SASPEQNQNHCSAINNSIPLMQGNLPY) from the N-terminal of human GnRH receptor

Molecular Weight: 54-60 kDa

Positive Control: Pituitary or normal ovary

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen GnRH Receptor / LH-RH Receptor Ab-3
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GNRH03; same as F1G4
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen A synthetic peptide aa 1-29 (MAN-SASPEQNQNHCSAINNSIPLMQGNLPY) from the N-terminal of human GnRH receptor
Quantity 1 mL
Regulatory Status RUO
Research Discipline G Protein-Coupled Receptors
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.