Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GnRH1 Monoclonal Antibody (LHRH13-387.5), Invitrogen™

Catalog No. PIMA546177
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546177 1 mg
PIMA546178 100 μg
01-671-844 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546177 Supplier Invitrogen™ Supplier No. MA546177
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10816 detects Gonadotropin Releasing Hormone from human samples. MA1-10816 has been successfully used in ELISA and immunohistochemistry applications. The MA1-10816 immunogen is purified human gonadotropin releasing hormone.

The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GnRH1
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone LHRH13-387.5
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene GNRH1
Gene Accession No. P01148
Gene Alias Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1
Gene Symbols GNRH1
Host Species Mouse
Immunogen Purified human gonadotropin releasing hormone.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2796
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.