Learn More
GnRH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA1120
Description
PA1-120 detects gonadotropin releasing hormone (GnRH) in mouse brain. PA1-120 has been successfully used in immunofluorescence and immunohistochemistry procedures. Immunofluorescent staining of GnRH in mouse embryo brain tissue using PA1-120 results in staining of neurons in the rostral forebrain. In immunohistochemical procedures, 4% paraformaldehyde is recommended as a fixative. The PA1-120 immunogen is a synthetic peptide corresponding to residues pyroE(22) H W S Y G L R P G-NH2(32) of mouse GnRH. This peptide (Cat. # PEP-168) is available for use in neutralization and control experiments.
GnRH1 Polyclonal antibody specifically detects GnRH1 in Mouse samples. It is validated for Immunohistochemistry.
Specifications
GnRH1 | |
Polyclonal | |
Unconjugated | |
GNRH1 | |
Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1 | |
Rabbit | |
Protein G | |
RUO | |
14714 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P13562 | |
GNRH1 | |
Synthetic peptide corresponding to residues pyroE(22) H W S Y G L R P G-NH2(32) of mouse GnRH. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.