Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GnRH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576386
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a secreted peptide hormone and member of the gonadotropin-releasing hormone (GnRH) family of proteins. The encoded protein regulates reproductive function by stimulating the production and release of the gonadotropins follicle-stimulating hormone (FSH) and luteinizing hormone (LH). The encoded protein may inhibit endometrial, ovarian, prostate, and breast cancer cell proliferation. Alternative splicing results in multiple transcript variants.
Specifications
GnRH2 | |
Polyclonal | |
Unconjugated | |
GNRH2 | |
GnRH II; GNRH2; GnRH-associated peptide 2; GnRH-associated peptide II; GnRH-II; Gonadoliberin II; Gonadoliberin-2; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 2; Gonadotropin-releasing hormone II; LH-RH II; LH-RHII; luliberin II; Luteinizing hormone-releasing hormone II; progonadoliberin II; Progonadoliberin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2797 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O43555 | |
GNRH2 | |
Recombinant protein of human GNRH2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction