Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GnRH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597047
Description
Immunogen sequence: QHWSHGWYPG GKRALSSAQD PQNALRPPGR ALDTAAGSPV QTAHGLPSDA LAPLDDSMPW EGRTTAQWSL HRKRHLARTL LTAAREPRPA PPSSNKV; Positive Samples: 293T.
This gene encodes a secreted peptide hormone and member of the gonadotropin-releasing hormone (GnRH) family of proteins. The encoded protein regulates reproductive function by stimulating the production and release of the gonadotropins follicle-stimulating hormone (FSH) and luteinizing hormone (LH). The encoded protein may inhibit endometrial, ovarian, prostate, and breast cancer cell proliferation. Alternative splicing results in multiple transcript variants.
Specifications
GnRH2 | |
Polyclonal | |
Unconjugated | |
GNRH2 | |
GnRH II; GNRH2; GnRH-associated peptide 2; GnRH-associated peptide II; GnRH-II; Gonadoliberin II; Gonadoliberin-2; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 2; Gonadotropin-releasing hormone II; LH-RH II; LH-RHII; luliberin II; Luteinizing hormone-releasing hormone II; progonadoliberin II; Progonadoliberin-2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2797 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.89 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43555 | |
GNRH2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 24-120 of human GNRH2 (NP_0014921). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction