Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GnRHR Antibody (GNRHR/2982R), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308770FR
Description
GnRHR Monoclonal antibody specifically detects GnRHR in Human samples. It is validated for ELISASpecifications
GnRHR | |
Monoclonal | |
DyLight 680 | |
GnRH receptor, GnRH-R, GNRHR1, gonadotropin-releasing hormone (type 1) receptor 1, gonadotropin-releasing hormone receptor, GRHR, leutinizing-releasing hormone receptor, LHRHRleutinizing hormone releasing horomone receptor, LRHR, luliberin receptor, type I GnRH receptor | |
Recombinant full-length human GnRHR protein | |
100 μg | |
GPCR | |
2798 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
GNRHR/2982R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction