Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GnRHR (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577448
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
This gene encodes the receptor for type 1 gonadotropin-releasing hormone. This receptor is a member of the seven-transmembrane, G-protein coupled receptor (GPCR) family. It is expressed on the surface of pituitary gonadotrope cells as well as lymphocytes, breast, ovary, and prostate. Following binding of gonadotropin-releasing hormone, the receptor associates with G-proteins that activate a phosphatidylinositol-calcium second messenger system. Activation of the receptor ultimately causes the release of gonadotropic luteinizing hormone (LH) and follicle stimulating hormone (FSH). Defects in this gene are a cause of hypogonadotropic hypogonadism (HH). Alternative splicing results in multiple transcript variants encoding different isoforms. More than 18 transcription initiation sites in the 5 region and multiple polyA signals in the 3 region have been identified for this gene.
Specifications
GnRHR (extracellular) | |
Polyclonal | |
Unconjugated | |
Gnrhr | |
Gonadotropin-releasing hormone receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14715, 81668 | |
-20°C | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.85 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P30969, Q01776 | |
Gnrhr | |
Peptide (C)NNASLEQDQNH, corresponding to amino acid residues 3-13 of rat GnRH Receptor | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction