Learn More
Invitrogen™ GnRHR Monoclonal Antibody (A9E4)

Description
A9E4 reacts with GnRH receptors in the anterior pituitary. GnRH stimulates the gonadotrophs of the anterior pituitary to secrete luteinising hormone (LH) as well as follicle-stimulating hormone (FSH). The receptor contains seven hydrophobic transmembrane domains connected by hydrophilic extracellular, and intracellular loops, characteristic of G-protein coupled receptors. Some cancers like ovarian and breast cancers sometimes carry GnRH receptors.
Specifications
Specifications
| Antigen | GnRHR |
| Applications | ELISA, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | A9E4 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Gnrhr |
| Gene Accession No. | P30968 |
| Gene Alias | GH1; GnRH receptor; Gnrhr; gnRH-R; GNRHR1; gonadotropin releasing hormone receptor; gonadotropin releasing hormone receptor type-I; gonadotropin-releasing hormone (type 1) receptor 1; gonadotropin-releasing hormone receptor; GRHR; growth hormone; HH7; HHG; hypergonadotropic hypogonadism; LCGR; leutinizing hormone releasing horomone receptor; leutinizing-releasing hormone receptor; LGR2; LH/CGR; LH/CG-R; LHCGR; LHR; Lh-rh receptor; Lhrhr; LRHR; LSH-R; luliberin receptor; Luteinizing hormone receptor; luteinizing hormone/choriogonadotropin receptor; lutropin/choriogonadotropin receptor; lutropin-choriogonadotropic hormone receptor; type I GnRH receptor; Type I GNRHR1; ULG5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.