Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GnRHR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103094
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103094 Supplier Invitrogen™ Supplier No. PA5103094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GNRHR.

This gene encodes the receptor for type 1 gonadotropin-releasing hormone. This receptor is a member of the seven-transmembrane, G-protein coupled receptor (GPCR) family. It is expressed on the surface of pituitary gonadotrope cells as well as lymphocytes, breast, ovary, and prostate. Following binding of gonadotropin-releasing hormone, the receptor associates with G-proteins that activate a phosphatidylinositol-calcium second messenger system. Activation of the receptor ultimately causes the release of gonadotropic luteinizing hormone (LH) and follicle stimulating hormone (FSH). Defects in this gene are a cause of hypogonadotropic hypogonadism (HH). Alternative splicing results in multiple transcript variants encoding different isoforms. More than 18 transcription initiation sites in the 5 region and multiple polyA signals in the 3 region have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GnRHR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gnrhr
Gene Accession No. P30968
Gene Alias GH1; GnRH receptor; Gnrhr; gnRH-R; GNRHR1; gonadotropin releasing hormone receptor; gonadotropin releasing hormone receptor type-I; gonadotropin-releasing hormone (type 1) receptor 1; gonadotropin-releasing hormone receptor; GRHR; growth hormone; HH7; HHG; hypergonadotropic hypogonadism; LCGR; leutinizing hormone releasing horomone receptor; leutinizing-releasing hormone receptor; LGR2; LH/CGR; LH/CG-R; LHCGR; LHR; Lh-rh receptor; Lhrhr; LRHR; LSH-R; luliberin receptor; Luteinizing hormone receptor; luteinizing hormone/choriogonadotropin receptor; lutropin/choriogonadotropin receptor; lutropin-choriogonadotropic hormone receptor; type I GnRH receptor; Type I GNRHR1; ULG5
Gene Symbols Gnrhr
Host Species Rabbit
Immunogen A synthesized peptide derived from human GNRHR(Accession P30968), corresponding to amino acid residues T42-L92.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2798
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.