Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GNRPX Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988654
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8654 Supplier Bioss Supplier No. BS8576R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen GNRPX
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PLEKHJ1
Gene Accession No. Q9NW61
Gene Alias 9530063M10Rik; Gnrpx; guanine nucleotide releasing protein x; Guanine nucleotide-releasing protein x; PH domain-containing family J member 1; pleckstrin homology domain containing J1; pleckstrin homology domain containing, family J member 1; Pleckstrin homology domain-containing family J member 1; Plekhj1
Gene Symbols PLEKHJ1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GNRPX/PLEKHJ1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55111
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.