Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GNS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110785
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110785 100 μL
PIPA5111276 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5110785 Supplier Invitrogen™ Supplier No. PA5110785
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVLVEYQGEG RNVTDPTCPS LSPGVSQCFP DCVCEDAYNN TYACVRTMSA LWNLQYCEFD DQEVFVEVYN LTADPDQITN IAKTIDPELL GKMNYRLMML QSCSGPTCRT PGVFDPGYRF DPRLMFSNRG SVRTRRF Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 93%.

GNS is a lysosomal enzyme found in all cells. It is involved in the catabolism of heparin, heparan sulphate, and keratan sulphate. Deficiency of this enzyme results in the accumulation of undegraded substrate and the lysosomal storage disorder mucopolysaccharidosis type IIID (Sanfilippo D syndrome). Mucopolysaccharidosis type IIID is the least common of the four subtypes of Sanfilippo syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GNS
Gene Accession No. P15586
Gene Alias 2610016K11Rik; AU042285; C87209; G6S; glucosamine (N-acetyl)-6-sulfatase; glucosamine (N-acetyl)-6-sulfatase (Sanfilippo disease IIID); glucosamine -6-sulfatase; glucosamine-6-sulfatase; Gns; N28088; N-acetylglucosamine 6-sulfatase precursor; N-acetylglucosamine-6-sulfatase
Gene Symbols GNS
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human GNS. Recombinant protein control fragment (Product #RP-90257).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.