Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Go 6983
GREENER_CHOICESDP

Catalog No. 22851 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-851 1 mg
22-851-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-851 Supplier Tocris Bioscience™ Supplier No. 2285/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Broad spectrum PKC inhibitor

Broad spectrum protein kinase C (PKC) inhibitor (IC50 values are 7, 7, 6, 10, 60 and 20000 nM for PKCα, PKCβ, PKCγ, PKCδ, PKCζ and PKCμ respectively). Displays cardioprotective properties; reduces polymorphonuclear leukocyte adherence and infiltration following myocardial ischemia/reperfusion injury. Optimizes naïve human pluripotent stem cell growth and viability following naïve cell derivation from primed ESCs and iPSCs using naïve human stem cell medium (NHSM).

Chemical Identifiers

CAS 133053-19-7
Molecular Formula C26H26N4O3

Specifications

Quantity 1 mg
Formula Weight 442.51
Purity >98%
Molecular Weight (g/mol) 442.519
Chemical Name or Material Go 6983
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.