Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ Go Direct™ Colorimeter
SDPSCI_ED

Catalog No. S39704 Shop All Vernier Software Products
Change view
Click to view available options
Range:
0 to 3 absorbance
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S39704 0 to 3 absorbance
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S39704 Supplier Vernier™ Supplier No. GDXCOL
Add to Cart
Add to Cart

Vernier™ Go Direct™ Colorimeter measures absorbance or transmittance of a liquid sample.

  • Useful range:
    • 0.05 to 1.0 (absorbance)
    • 90% to 10% transmittance (%T)
  • Wavelengths: 430 nm, 470 nm, 565 nm, 635 nm
  • Conduct Beer's law investigations.
  • Determine the concentration of unknown solutions.
  • Study changes in concentration vs. time.
  • Monitor reaction rates.

Specifications

Grade Middle School, High School, College
Interface Bluetooth or USB
Warranty 5 Year Limited
For Use With (Application) Graphical Analysis 4 or LabQuest App
Range 0 to 3 absorbance
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.