Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GO Protein alpha Antibody, Novus Biologicals™
SDP

Catalog No. NB017772 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017772 0.1 mL
NB10066416 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017772 Supplier Novus Biologicals Supplier No. NB100664160.1ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GO Protein alpha Polyclonal specifically detects GO Protein alpha in Mouse, Drosophila samples. It is validated for Western Blot, ELISA.

Specifications

Antigen GO Protein alpha
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:500-1:1000
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Gene Accession No. P18872
Gene Alias DKFZp686O0962, G-ALPHA-o, GNAO, guanine nucleotide binding protein (G protein), alpha activating activitypolypeptide O, guanine nucleotide binding protein, alpha activating polypeptide O, guanine nucleotide-binding protein G(o) subunit alpha
Gene Symbols GNAO1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the amino acids 345 - 354 of the native molecule conjugated to Keyhole Limpet Haemocyanin.
Molecular Weight of Antigen 40 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2775
Test Specificity G PROTEIN ALPHA 0
Target Species Drosophila, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.