Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

13(S)-HODE, Goat, Polyclonal Antibody, Abnova™

Catalog No. 89137634 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-634 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-634 Supplier Abnova Corporation Supplier No. PAB13604
Add to Cart
Add to Cart

Goat polyclonal antibody raised against 13(S)-HODE.

Specifications

Antigen 13(S)-HODE
Applications ELISA, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Goat polyclonal antibody raised against 13(S)-HODE.
Dilution Immunohistochemistry (2 ug/well) Enzyme Immunoassay (2 ug/well) Radioimmunoassay (2 ug/well) The optimal working dilution should be determined by the end user.
Host Species Goat
Immunogen 13(S)-HODE.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity 13(S)-HODE 100.0 % 13(R)-HODE 11.7 % 9-oxo-ODE 11.7 % 15(S)-HETE 6.0 % 11(S)-HETE 2.7 % 9(R)-Hode 0.8 % 9(s)-HODE 0.6 % Linoleic Acid 0.5 % 13-oxo-ODE 0.4 %
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.