Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ABCC4 Goat anti-Human, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89132611 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-132-611 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-132-611 Supplier Abnova Corporation Supplier No. PAB6698
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of ABCC4.

The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily which is involved in multi-drug resistance. The specific function of this protein has not yet been determined; however, this protein may play a role in cellular detoxification as a pump for its substrate, organic anions. Alternative splicing results in multiple splice variants encoding different isoforms. [provided by RefSeq

Sequence: CNGQPSTLTIFETAL

Specifications

Antigen ABCC4
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of ABCC4.
Dilution ELISA (1:32000) Western Blot (1-3 ug/mL) Immunohistochemistry (2-4 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
Gene ABCC4
Gene Accession No. NP_005836.2
Gene Alias EST170205/MOAT-B/MOATB/MRP4
Gene Symbols ABCC4
Host Species Goat
Immunogen A synthetic peptide corresponding to C-terminus of human ABCC4.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10257
Test Specificity This antibody is expected to recognize isoform 1 (NP_005836.2) only.
Target Species Human, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.