Learn More
ACADVL, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ACADVL.
Supplier: Abnova Corporation PAB18538
Description
The protein encoded by this gene is targeted to the inner mitochondrial membrane where it catalyzes the first step of the mitochondrial fatty acid beta-oxidation pathway. This acyl-Coenzyme A dehydrogenase is specific to long-chain and very-long-chain fatty acids. A deficiency in this gene product reduces myocardial fatty acid beta-oxidation and is associated with cardiomyopathy. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq
Sequence: DKSDSHPSDALTRK-CSpecifications
ACADVL | |
Polyclonal | |
Unconjugated | |
ELISA (1:2000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
ACADVL | |
ACAD6/LCACD/VLCAD | |
Goat | |
Antigen affinity purification | |
RUO | |
37 | |
Human | |
Liquid |
ELISA, Western Blot | |
0.5 mg/mL | |
Goat polyclonal antibody raised against synthetic peptide of ACADVL. | |
In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
NP_000009.1;NP_001029031.1 | |
ACADVL | |
A synthetic peptide corresponding to amino acids near N-terminus of human ACADVL. | |
100 μg | |
Primary | |
This antibody is expected to recognize both reported isoforms (NP_000009.1; NP_001029031.1). | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only