Learn More
ACPP Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ACPP.
Supplier: Abnova Corporation PAB19474
Description
This gene encodes an enzyme that catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is synthesized under androgen regulation and is secreted by the epithelial cells of the prostate gland. An alternatively spliced transcript variant encoding a longer isoform has been found for this gene. This isoform contains a transmembrane domain and is localized in the plasma membrane-endosomal-lysosomal pathway. [provided by RefSeq
Sequence: C-NHMKRATQIPSYKKSpecifications
ACPP | |
Polyclonal | |
Goat polyclonal antibody raised against synthetic peptide of ACPP. | |
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
NP_001127666.1;NP_001090.2 | |
ACPP | |
A synthetic peptide corresponding to internal region of human ACPP. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:3200) Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user. | |
ACPP | |
ACP-3/ACP3/PAP | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
55 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only