Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADAMTS8 Goat, Polyclonal Antibody, Abnova™

Catalog No. 89120164 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-164 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-164 Supplier Abnova Corporation Supplier No. PAB18607
Add to Cart
Add to Cart

Goat polyclonal antibody raised against synthetic peptide of ADAMTS8.

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains two C-terminal TS motifs, and disrupts angiogenesis in vivo. A number of disorders have been mapped in the vicinity of this gene, most notably lung neoplasms. [provided by RefSeq

Sequence: C-RQERGDHQED

Specifications

Antigen ADAMTS8
Applications ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Description Goat polyclonal antibody raised against synthetic peptide of ADAMTS8.
Dilution ELISA (1:4000) The optimal working dilution should be determined by the end user.
Formulation In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA)
Gene ADAMTS8
Gene Accession No. NP_008968.4
Gene Alias ADAM-TS8/FLJ41712/METH2
Gene Symbols ADAMTS8
Host Species Goat
Immunogen A synthetic peptide corresponding to amino acids 176-185 at internal region of human ADAMTS8.
Purification Method Antigen affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11095
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.